Anti-CD4 Rabbit Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB4830-100EA 2330 DKK
BIRBORB4830-100 BIRBORB4830-200
Anti-CD4 Rabbit Polyclonal Antibody
Antistoffer
Immunochemistry: 100-1000; Immunofluorescence: 1:200

Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR