Anti-MGRN1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB3728-100EA 0 DKK
BIRBORB3728-100
Anti-MGRN1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Primary
Antigen: MGRN1 (mahogunin ring finger 1, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR