Anti-IL31RA Mouse Monoclonal Antibody [clone: 3A10]

Leverandør: Abnova
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABNOH00133396-M01EA 2940 DKK
ABNOH00133396-M01
Anti-IL31RA Mouse Monoclonal Antibody [clone: 3A10]
Antistoffer
Type: Primary
Antigen: IL31RA
Clonality: Monoclonal
Clone: 3A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR