Anti-COLEC10 Mouse Monoclonal Antibody [clone: 1D11]
Leverandør: Abnova
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABNOH00010584-M02EA 2940 DKK
ABNOH00010584-M02
Anti-COLEC10 Mouse Monoclonal Antibody [clone: 1D11]
Antistoffer
Type: Primary
Antigen: COLEC10
Clonality: Monoclonal
Clone: 1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human