Anti-AGPAT1 Mouse Polyclonal Antibody
Leverandør: Abnova
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABNOH00010554-B01PEA 2940 DKK
ABNOH00010554-B01P
Anti-AGPAT1 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: AGPAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human