Anti-SRI Mouse Monoclonal Antibody [clone: 1E12]

Leverandør: Abnova
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABNOH00006717-M01EA 2940 DKK
ABNOH00006717-M01
Anti-SRI Mouse Monoclonal Antibody [clone: 1E12]
Antistoffer
Type: Primary
Antigen: SRI
Clonality: Monoclonal
Clone: 1E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR