Anti-SERPINC1 Mouse Monoclonal Antibody [clone: 2B12]

Leverandør: Abnova
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABNOH00000462-M02EA 2940 DKK
ABNOH00000462-M02
Anti-SERPINC1 Mouse Monoclonal Antibody [clone: 2B12]
Antistoffer
Type: Primary
Antigen: SERPINC1
Clonality: Monoclonal
Clone: 2B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR