Anti-TMEFF2 Mouse Monoclonal Antibody [clone: OTI3D1]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA504464EA 4600 DKK
ORIGTA504464
Anti-TMEFF2 Mouse Monoclonal Antibody [clone: OTI3D1]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: TMEFF2 (transmembrane protein with EGF-like and two follistatin-like domains 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR