Anti-PSMB7 Mouse Monoclonal Antibody [clone: OTI1B2]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA504456EA 4340 DKK
ORIGTA504456
Anti-PSMB7 Mouse Monoclonal Antibody [clone: OTI1B2]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1000

Type: Primary
Antigen: PSMB7 (proteasome (prosome, macropain) subunit, beta type, 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR