Anti-EXOSC7 Mouse Monoclonal Antibody [clone: OTI1F7]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA504229EA 4500 DKK
ORIGTA504229
Anti-EXOSC7 Mouse Monoclonal Antibody [clone: OTI1F7]
Antistoffer
  • Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: EXOSC7 (exosome component 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR