Anti-GSTO2 Mouse Monoclonal Antibody [clone: OTI3A9]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA504087EA 4600 DKK
ORIGTA504087
Anti-GSTO2 Mouse Monoclonal Antibody [clone: OTI3A9]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: GSTO2 (glutathione S-transferase omega 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR