Anti-USP7 Mouse Monoclonal Antibody [clone: OTI3D3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA504060EA 4500 DKK
ORIGTA504060
Anti-USP7 Mouse Monoclonal Antibody [clone: OTI3D3]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: USP7 (ubiquitin specific peptidase 7 (herpes virus-associated))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR