Anti-SIX1 Mouse Monoclonal Antibody [clone: OTI1H4]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA504057EA 4540 DKK
ORIGTA504057
Anti-SIX1 Mouse Monoclonal Antibody [clone: OTI1H4]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SIX1 (SIX homeobox 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR