Anti-TIMP2 Mouse Monoclonal Antibody [clone: OTI2G7]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA504018EA 4600 DKK
ORIGTA504018
Anti-TIMP2 Mouse Monoclonal Antibody [clone: OTI2G7]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: TIMP2 (TIMP metallopeptidase inhibitor 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR