Anti-NAPEPLD Mouse Monoclonal Antibody [clone: OTI5F7]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA503860EA 4640 DKK
ORIGTA503860
Anti-NAPEPLD Mouse Monoclonal Antibody [clone: OTI5F7]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NAPEPLD (N-acyl phosphatidylethanolamine phospholipase D)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR