Anti-LPL Mouse Monoclonal Antibody [clone: OTI4H3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA503787EA 4500 DKK
ORIGTA503787
Anti-LPL Mouse Monoclonal Antibody [clone: OTI4H3]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: LPL (lipoprotein lipase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR