Anti-METTL21A Mouse Monoclonal Antibody [clone: OTI2D6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA503703EA 4500 DKK
ORIGTA503703
Anti-METTL21A Mouse Monoclonal Antibody [clone: OTI2D6]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: METTL21A (methyltransferase like 21A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR