Anti-DPP10 Mouse Monoclonal Antibody [clone: OTI1A5]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA503585EA 4600 DKK
ORIGTA503585
Anti-DPP10 Mouse Monoclonal Antibody [clone: OTI1A5]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: DPP10 (dipeptidyl-peptidase 10 (non-functional))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR