Anti-EIF2B3 Mouse Monoclonal Antibody [clone: OTI1H3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA503562EA 4500 DKK
ORIGTA503562
Anti-EIF2B3 Mouse Monoclonal Antibody [clone: OTI1H3]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: EIF2B3 (eukaryotic translation initiation factor 2B, subunit 3 gamma, 58kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR