Anti-PDE4B Mouse Monoclonal Antibody [clone: OTI3B12]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA503477EA 4500 DKK
ORIGTA503477
Anti-PDE4B Mouse Monoclonal Antibody [clone: OTI3B12]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: PDE4B (phosphodiesterase 4B, cAMP-specific)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR