Anti-PGAM2 Mouse Monoclonal Antibody [clone: OTI1C11]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA503403EA 4600 DKK
ORIGTA503403
Anti-PGAM2 Mouse Monoclonal Antibody [clone: OTI1C11]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PGAM2 (phosphoglycerate mutase 2 (muscle))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR