Anti-CYB5R2 Mouse Monoclonal Antibody [clone: OTI4B9]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA503251EA 4470 DKK
ORIGTA503251
Anti-CYB5R2 Mouse Monoclonal Antibody [clone: OTI4B9]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: CYB5R2 (cytochrome b5 reductase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR