Anti-GATM Mouse Monoclonal Antibody [clone: OTI1C9]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGTA503147EA 4500 DKK
ORIGTA503147
Anti-GATM Mouse Monoclonal Antibody [clone: OTI1C9]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: GATM (glycine amidinotransferase (L-arginine:glycine amidinotransferase))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human