Anti-MGRN1 Mouse Monoclonal Antibody [clone: OTI3A1]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA502681EA 4430 DKK
ORIGTA502681
Anti-MGRN1 Mouse Monoclonal Antibody [clone: OTI3A1]
Antistoffer
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MGRN1 (mahogunin ring finger 1, E3 ubiquitin protein ligase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR