Anti-CHCHD5 Mouse Monoclonal Antibody [clone: OTI9E11]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA502668EA 4540 DKK
ORIGTA502668
Anti-CHCHD5 Mouse Monoclonal Antibody [clone: OTI9E11]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: CHCHD5 (coiled-coil-helix-coiled-coil-helix domain containing 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR