Anti-KCTD14 Mouse Monoclonal Antibody [clone: OTI1A11]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA502637EA 4340 DKK
ORIGTA502637
Anti-KCTD14 Mouse Monoclonal Antibody [clone: OTI1A11]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: KCTD14 (potassium channel tetramerization domain containing 14)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR