Anti-LACTB2 Mouse Monoclonal Antibody [clone: OTI3A5]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA502580EA 4340 DKK
ORIGTA502580
Anti-LACTB2 Mouse Monoclonal Antibody [clone: OTI3A5]
Antistoffer
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: LACTB2 (lactamase, beta 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR