Anti-BTRC Mouse Monoclonal Antibody [clone: OTI1A4]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA502524EA 4600 DKK
ORIGTA502524
Anti-BTRC Mouse Monoclonal Antibody [clone: OTI1A4]
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: BTRC (beta-transducin repeat containing E3 ubiquitin protein ligase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR