Anti-ZFAND2B Mouse Monoclonal Antibody [clone: OTI2G10]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA502514EA 4430 DKK
ORIGTA502514
Anti-ZFAND2B Mouse Monoclonal Antibody [clone: OTI2G10]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: ZFAND2B (zinc finger, AN1-type domain 2B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR