Anti-RASSF1 Mouse Monoclonal Antibody [clone: OTI8E2]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA502486EA 4600 DKK
ORIGTA502486
Anti-RASSF1 Mouse Monoclonal Antibody [clone: OTI8E2]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: RASSF1 (Ras association (RalGDS/AF-6) domain family member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR