Anti-EIF4E2 Mouse Monoclonal Antibody [clone: OTI1F11]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA502403EA 4540 DKK
ORIGTA502403
Anti-EIF4E2 Mouse Monoclonal Antibody [clone: OTI1F11]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: EIF4E2 (eukaryotic translation initiation factor 4E family member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR