Anti-SNTA1 Mouse Monoclonal Antibody [clone: OTI2D5]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA502041EA 4500 DKK
ORIGTA502041
Anti-SNTA1 Mouse Monoclonal Antibody [clone: OTI2D5]
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SNTA1 (syntrophin, alpha 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR