Anti-NHEJ1 Mouse Monoclonal Antibody [clone: OTI3E6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA502004EA 4340 DKK
ORIGTA502004
Anti-NHEJ1 Mouse Monoclonal Antibody [clone: OTI3E6]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NHEJ1 (nonhomologous end-joining factor 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR