Anti-PBLD Mouse Monoclonal Antibody [clone: OTI7G5]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGTA501994EA 4540 DKK
ORIGTA501994
Anti-PBLD Mouse Monoclonal Antibody [clone: OTI7G5]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: PBLD (phenazine biosynthesis-like protein domain containing)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human