Anti-GSTA4 Mouse Monoclonal Antibody [clone: OTI3F5]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA501903EA 4540 DKK
ORIGTA501903
Anti-GSTA4 Mouse Monoclonal Antibody [clone: OTI3F5]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1:2000; Flow Cytometry: 1:100; Immunochemistry: 1:50-1:150

Type: Primary
Antigen: GSTA4 (glutathione S-transferase alpha 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR