Anti-AK3 Mouse Monoclonal Antibody [clone: OTI6D6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA501833EA 4640 DKK
ORIGTA501833
Anti-AK3 Mouse Monoclonal Antibody [clone: OTI6D6]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: AK3 (adenylate kinase 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR