Anti-AFP Mouse Monoclonal Antibody [clone: OTI2A9]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA501782EA 4500 DKK
ORIGTA501782
Anti-AFP Mouse Monoclonal Antibody [clone: OTI2A9]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: AFP (alpha-fetoprotein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR