Anti-MTMR14 Mouse Monoclonal Antibody [clone: OTI6B6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA501774EA 4380 DKK
ORIGTA501774
Anti-MTMR14 Mouse Monoclonal Antibody [clone: OTI6B6]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MTMR14 (myotubularin related protein 14)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR