Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: OTI7G9]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA501762EA 4430 DKK
ORIGTA501762
Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: OTI7G9]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PIK3AP1 (phosphoinositide-3-kinase adaptor protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR