Anti-SULT2A1 Mouse Monoclonal Antibody [clone: OTI3E8]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA501657EA 4600 DKK
ORIGTA501657
Anti-SULT2A1 Mouse Monoclonal Antibody [clone: OTI3E8]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: SULT2A1 (sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA)-preferring, member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR