Anti-NSMCE2 Mouse Monoclonal Antibody [clone: OTI6A6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA501635EA 4540 DKK
ORIGTA501635
Anti-NSMCE2 Mouse Monoclonal Antibody [clone: OTI6A6]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NSMCE2 (non-SMC element 2, MMS21 homolog (S. cerevisiae))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR