Anti-EPHX2 Mouse Monoclonal Antibody [clone: OTI3F6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA501628EA 4500 DKK
ORIGTA501628
Anti-EPHX2 Mouse Monoclonal Antibody [clone: OTI3F6]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: EPHX2 (epoxide hydrolase 2, cytoplasmic)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR