Anti-PEBP1 Mouse Monoclonal Antibody [clone: OTI2D4]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA501616EA 4640 DKK
ORIGTA501616
Anti-PEBP1 Mouse Monoclonal Antibody [clone: OTI2D4]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PEBP1 (phosphatidylethanolamine binding protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR