Anti-FSHB Mouse Monoclonal Antibody [clone: OTI1B2]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA501602EA 4430 DKK
ORIGTA501602
Anti-FSHB Mouse Monoclonal Antibody [clone: OTI1B2]
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: FSHB (follicle stimulating hormone, beta polypeptide)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR