Anti-MPP3 Mouse Monoclonal Antibody [clone: OTI2A11]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA501564EA 4340 DKK
ORIGTA501564
Anti-MPP3 Mouse Monoclonal Antibody [clone: OTI2A11]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MPP3 (membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR