Anti-XPNPEP1 Mouse Monoclonal Antibody [clone: OTI1E3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA501383EA 4540 DKK
ORIGTA501383
Anti-XPNPEP1 Mouse Monoclonal Antibody [clone: OTI1E3]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: XPNPEP1 (X-prolyl aminopeptidase (aminopeptidase P) 1, soluble)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR