Anti-C21orf59 Mouse Monoclonal Antibody [clone: OTI1A1]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA501360EA 4500 DKK
ORIGTA501360
Anti-C21orf59 Mouse Monoclonal Antibody [clone: OTI1A1]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: C21orf59 (chromosome 21 open reading frame 59)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR