Anti-ARNTL Mouse Monoclonal Antibody [clone: OTI1C11]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA501174EA 4500 DKK
ORIGTA501174
Anti-ARNTL Mouse Monoclonal Antibody [clone: OTI1C11]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: ARNTL (aryl hydrocarbon receptor nuclear translocator-like)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR