Anti-DNTTIP1 Mouse Monoclonal Antibody [clone: OTI1D3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA501170EA 4600 DKK
ORIGTA501170
Anti-DNTTIP1 Mouse Monoclonal Antibody [clone: OTI1D3]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: DNTTIP1 (deoxynucleotidyltransferase, terminal, interacting protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR