Anti-SRR Mouse Monoclonal Antibody [clone: OTI7G6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA500958EA 4600 DKK
ORIGTA500958
Anti-SRR Mouse Monoclonal Antibody [clone: OTI7G6]
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: SRR (serine racemase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR